Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM17
Entrez Gene ID
694286
Gene Name
tripartite motif containing 17
Gene Aliases
-
Chromosome Location
Chr.1: 85984529 - 85993337 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082856.4 XP_001082856.1 - 1410 135
XM_015114701.2 XP_014970187.1 - 1267 135

View Full Product Details