Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEPRO
Entrez Gene ID
708933
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
C3orf17
Chromosome Location
Chr.2: 163210416 - 163228024 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105980.4 XP_001105980.3 - 1159 152
XM_002802647.3 XP_002802693.3 - 962 152
XM_002802648.3 XP_002802694.3 - 1026 152
XM_015129867.2 XP_014985353.2 - 944 152
XM_015129869.2 XP_014985355.2 - 950 152

View Full Product Details