Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGS7BP
Entrez Gene ID
695221
Gene Name
regulator of G protein signaling 7 binding protein
Gene Aliases
EGK_16530
Chromosome Location
Chr.6: 63029794 - 63137597 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266063.1 NP_001252992.1 - 467 147
XM_015140026.2 XP_014995512.1 - 951 147
GenBank mRNA JU334488.1 - - 467 147
JV683952.1 - - 469 147

View Full Product Details