Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COX19
Entrez Gene ID
696368
Gene Name
COX19 cytochrome c oxidase assembly homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.3: 34045464 - 34052598 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257834.1 NP_001244763.1 - 176 184
GenBank mRNA JU323489.1 - - 176 184
JU477741.1 - - 113 184
JV048636.1 - - 106 184

View Full Product Details