Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPN20
Entrez Gene ID
711276
Gene Name
protein tyrosine phosphatase non-receptor type 20
Gene Aliases
EGK_19635, PTPN20A
Chromosome Location
Chr.9: 49239796 - 49340960 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109336.4 XP_001109336.1 - 738 104
XM_015146962.2 XP_015002448.1 - 1226 104
XM_015146965.2 XP_015002451.1 - 2484 104
XM_015146968.2 XP_015002454.1 - 648 104
XM_015146970.2 XP_015002456.1 - 559 104
XM_015146971.2 XP_015002457.1 - 470 104
XM_028826354.1 XP_028682187.1 - 1188 104
XM_028826355.1 XP_028682188.1 - 1289 104
XM_028826356.1 XP_028682189.1 - 791 104
XM_028826357.1 XP_028682190.1 - 1096 104

View Full Product Details