Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGRP
Entrez Gene ID
698479
Gene Name
agouti related neuropeptide
Gene Aliases
EGK_12895
Chromosome Location
Chr.20: 54671685 - 54673202 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028840938.1 XP_028696771.1 - 918 132

View Full Product Details