Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSV
Entrez Gene ID
710850
Gene Name
cathepsin V
Gene Aliases
CTSL2
Chromosome Location
Chr.15: 110985513 - 111007231 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106814.4 XP_001106814.3 - 1026 119
XM_015118134.2 XP_014973620.2 - 980 119
XM_028835374.1 XP_028691207.1 - 914 119
XM_028835375.1 XP_028691208.1 - 920 119
XM_028835376.1 XP_028691209.1 - 978 119
XM_028835377.1 XP_028691210.1 - 927 119
GenBank mRNA JU323374.1 - - 840 119
JU481076.1 - - 777 119
JV044398.1 - - 813 119
JV637693.1 - - 840 119

View Full Product Details