Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMP1
Entrez Gene ID
698319
Gene Name
epithelial membrane protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 14587302 - 14607648 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087779.4 XP_001087779.2 - 709 175

View Full Product Details