Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP4
Entrez Gene ID
701365
Gene Name
fatty acid binding protein 4
Gene Aliases
EGK_19076
Chromosome Location
Chr.8: 81955619 - 81960248 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266067.1 NP_001252996.1 - 295 162
GenBank mRNA JU473048.1 - - 295 162

View Full Product Details