Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLOC1S1
Entrez Gene ID
707395
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 1
Gene Aliases
-
Chromosome Location
Chr.11: 55275769 - 55279465 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015151916.2 XP_015007402.2 - 396 73
GenBank mRNA JU472985.1 - - 353 73
JV491096.1 - - 362 73

View Full Product Details