Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PITX1
Entrez Gene ID
711666
Gene Name
paired like homeodomain 1
Gene Aliases
-
Chromosome Location
Chr.6: 132486974 - 132493513 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028849736.1 XP_028705569.1 - 548 132

View Full Product Details