Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHEX
Entrez Gene ID
699012
Gene Name
hematopoietically expressed homeobox
Gene Aliases
-
Chromosome Location
Chr.9: 92854406 - 92860107 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001090715.4 XP_001090715.2 - 660 129
GenBank mRNA JU326604.1 - - 105 129
JU485368.1 - - 175 129
JV713395.1 - - 373 129

View Full Product Details