The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257799.1 | NP_001244728.1 | - | 684 | 149 |
XM_015109888.2 | XP_014965374.1 | - | 760 | 149 | |
XM_028830408.1 | XP_028686241.1 | - | 933 | 149 | |
XM_028830409.1 | XP_028686242.1 | - | 665 | 149 | |
GenBank mRNA | JU322309.1 | - | - | 684 | 149 |
JU472113.1 | - | - | 683 | 149 | |
JV043915.1 | - | - | 682 | 149 |