Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCVRN
Entrez Gene ID
717807
Gene Name
recoverin
Gene Aliases
EGK_08191
Chromosome Location
Chr.16: 9689117 - 9698615 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194602.2 NP_001181531.1 - 495 109
GenBank mRNA JU322773.1 - - 512 109
JV043451.1 - - 564 109
JV567850.1 - - 539 109

View Full Product Details