Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- XCL1
- Entrez Gene ID
-
574367
- Gene Name
- chemokine (C motif) ligand 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 25890736 - 25893721 on Build Mmul_10
- Gene Symbol
- LOC100423131
- Entrez Gene ID
-
100423131
- Gene Name
- lymphotactin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 25708329 - 25932829 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV554243.1 |
- |
- |
78 |
104 |
View Full Product Details