Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK1
Entrez Gene ID
708951
Gene Name
serine peptidase inhibitor Kazal type 1
Gene Aliases
EGK_16991
Chromosome Location
Chr.6: 145242056 - 145249152 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028849824.1 XP_028705657.1 - 308 98

View Full Product Details