Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STC1
Entrez Gene ID
710782
Gene Name
stanniocalcin 1
Gene Aliases
EGK_18778
Chromosome Location
Chr.8: 24104090 - 24115266 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261309.1 NP_001248238.1 - 310 100
GenBank mRNA JV048053.1 - - 310 100
JV659730.1 - - 318 100

View Full Product Details