Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZNF84
- Entrez Gene ID
-
694939
- Gene Name
- zinc finger protein 84
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 132840490 - 132862690 on Build Mmul_10
- Gene Symbol
- ZFP90
- Entrez Gene ID
-
704279
- Gene Name
- ZFP90 zinc finger protein
- Gene Aliases
-
EGK_12927
- Chromosome Location
-
Chr.20: 55771154 - 55807894 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV673788.1 |
- |
- |
248 |
88 |
View Full Product Details