Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FZD6
Entrez Gene ID
694625
Gene Name
frizzled class receptor 6
Gene Aliases
EGK_19188, Frizzled-6
Chromosome Location
Chr.8: 103575786 - 103609520 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265729.1 NP_001252658.1 - 378 146
XM_015145733.2 XP_015001219.1 - 426 146
GenBank mRNA JV072749.1 - - 414 146
JV636398.1 - - 378 146

View Full Product Details