Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAGLN2
Entrez Gene ID
719527
Gene Name
transgelin 2
Gene Aliases
-
Chromosome Location
Chr.1: 91017933 - 91025320 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265900.1 NP_001252829.1 - 422 106
XM_015110251.2 XP_014965737.1 - 509 106
XM_015110257.2 XP_014965743.1 - 512 106
XM_028846138.1 XP_028701971.1 - 386 106
XM_028846145.1 XP_028701978.1 - 386 106
XM_028846149.1 XP_028701982.1 - 386 106
XM_028846153.1 XP_028701986.1 - 404 106
GenBank mRNA JU336656.1 - - 416 106
JU477453.1 - - 388 106
JV048413.1 - - 422 106
JV663133.1 - - 426 106

View Full Product Details