Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STC2
Entrez Gene ID
703900
Gene Name
stanniocalcin 2
Gene Aliases
EGK_17143
Chromosome Location
Chr.6: 170721088 - 170735923 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097342.4 XP_001097342.1 - 1877 102

View Full Product Details