Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSRP1
Entrez Gene ID
709693
Gene Name
cysteine and glycine rich protein 1
Gene Aliases
EGK_05263
Chromosome Location
Chr.1: 58904700 - 58967499 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU323705.1 - - 156 114
JU474399.1 - - 80 114
JV673835.1 - - 70 114

View Full Product Details