Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9A3R1
Entrez Gene ID
703933
Gene Name
SLC9A3 regulator 1
Gene Aliases
-
Chromosome Location
Chr.16: 71087630 - 71106727 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266178.1 NP_001253107.1 - 777 64
GenBank mRNA JU322781.1 - - 777 64
JU496501.1 - - 812 64

View Full Product Details