Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A13
Entrez Gene ID
696240
Gene Name
solute carrier family 22 member 13
Gene Aliases
EGK_11793
Chromosome Location
Chr.2: 93688278 - 93700692 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087330.4 XP_001087330.1 - 740 121

View Full Product Details