Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGAP35
Entrez Gene ID
716936
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRLF1
Chromosome Location
Chr.19: 46862330 - 47013582 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028839122.1 XP_028694955.1 - 4201 73
GenBank mRNA JU334158.1 - - 3683 73
JU475285.1 - - 3690 73
JV046843.1 - - 3688 73

View Full Product Details