Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH7
Entrez Gene ID
699343
Gene Name
alkB homolog 7
Gene Aliases
-
Chromosome Location
Chr.19: 6305373 - 6308523 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087443.4 XP_001087443.2 - 1280 131
GenBank mRNA JU336690.1 - - 614 131
JU477484.1 - - 615 131

View Full Product Details