Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD5
Entrez Gene ID
699323
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
-
Chromosome Location
Chr.13: 15753710 - 15758408 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088583.4 XP_001088583.2 - 355 79
GenBank mRNA JU336691.1 - - 240 79
JU477485.1 - - 267 79
JV048433.1 - - 224 79
JV663335.1 - - 240 79

View Full Product Details