Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD6
Entrez Gene ID
716767
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
-
Chromosome Location
Chr.2: 149548548 - 149787141 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266664.1 NP_001253593.1 - 730 132
XM_015129926.2 XP_014985412.2 - 1190 132
XM_015129927.2 XP_014985413.2 - 1187 132
XM_015129928.2 XP_014985414.2 - 1396 132
XM_015129932.2 XP_014985418.2 - 866 132
XM_015129933.2 XP_014985419.1 - 799 132
XM_028843420.1 XP_028699253.1 - 1289 132
XM_028843421.1 XP_028699254.1 - 1001 132
GenBank mRNA JU336706.1 - - 732 132
JV048439.1 - - 730 132
JV637257.1 - - 728 132
JV698622.1 - - 740 132

View Full Product Details