Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEDAG
Entrez Gene ID
721932
Gene Name
mesenteric estrogen dependent adipogenesis
Gene Aliases
-
Chromosome Location
Chr.17: 9955882 - 9975265 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015120827.2 XP_014976313.1 - 836 75
GenBank mRNA JV725557.1 - - 219 75

View Full Product Details