Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM5
Entrez Gene ID
574288
Gene Name
tripartite motif containing 5
Gene Aliases
-
Chromosome Location
Chr.14: 62539688 - 62568289 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032910.1 NP_001028082.1 - 212 77
GenBank mRNA AY523632.1 - - 212 77

View Full Product Details