Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNSTRN
Entrez Gene ID
702753
Gene Name
kinetochore localized astrin (SPAG5) binding protein
Gene Aliases
C7H15orf23
Chromosome Location
Chr.7: 16792513 - 16804120 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015141933.2 XP_014997419.2 - 557 111
XM_028850740.1 XP_028706573.1 - 559 111
XM_028850741.1 XP_028706574.1 - 561 111
GenBank mRNA JU495945.1 - - 287 111
JV075023.1 - - 291 111

View Full Product Details