Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCMTD1
Entrez Gene ID
717131
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases
EGK_18924
Chromosome Location
Chr.8: 52417773 - 52496859 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260718.1 NP_001247647.1 - 972 96
XM_015145337.2 XP_015000823.1 - 993 96
XM_015145338.2 XP_015000824.1 - 633 96
XM_015145339.2 XP_015000825.1 - 848 96
XM_015145341.2 XP_015000827.1 - 498 96
GenBank mRNA JU322194.1 - - 957 96
JU471616.1 - - 930 96
JV044653.1 - - 972 96

View Full Product Details