Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP3A43
Entrez Gene ID
709770
Gene Name
cytochrome P450, family 3, subfamily A, polypeptide 43
Gene Aliases
-
Chromosome Location
Chr.3: 41999570 - 42044248 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127630.1 NP_001121102.1 - 278 86
XM_028844981.1 XP_028700814.1 - 528 86
XM_028844982.1 XP_028700815.1 - 2698 86
XM_028844983.1 XP_028700816.1 - 2794 86
XM_028844984.1 XP_028700817.1 - 160 86
XM_028844985.1 XP_028700818.1 - 466 86
GenBank mRNA EF589791.1 - - 425 86
EF589792.1 - - 278 86

View Full Product Details