Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPH6
Entrez Gene ID
701401
Gene Name
diphthamine biosynthesis 6
Gene Aliases
ATPBD4
Chromosome Location
Chr.7: 11698836 - 11998199 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266595.1 NP_001253524.1 - 314 91
XM_028850278.1 XP_028706111.1 - 392 91
GenBank mRNA JU336983.1 - - 336 91
JU477708.1 - - 314 91

View Full Product Details