Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSAD2
Entrez Gene ID
708720
Gene Name
radical S-adenosyl methionine domain containing 2
Gene Aliases
-
Chromosome Location
Chr.13: 101877710 - 101901657 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281312.1 NP_001268241.1 - 342 117
GenBank mRNA JQ437830.1 - - 342 117

View Full Product Details