Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPHA2
Entrez Gene ID
717261
Gene Name
glycoprotein hormone subunit alpha 2
Gene Aliases
-
Chromosome Location
Chr.14: 9598334 - 9601344 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194654.1 NP_001181583.1 - 153 64
XM_028832810.1 XP_028688643.1 - 365 64

View Full Product Details