Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A46
Entrez Gene ID
705968
Gene Name
solute carrier family 25 member 46
Gene Aliases
-
Chromosome Location
Chr.6: 108172608 - 108199750 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266009.2 NP_001252938.1 - 452 125
GenBank mRNA JU337036.1 - - 455 125
JU477768.1 - - 457 125
JV045151.1 - - 452 125

View Full Product Details