Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYPLAL1
Entrez Gene ID
707313
Gene Name
lysophospholipase like 1
Gene Aliases
-
Chromosome Location
Chr.1: 76703980 - 76742960 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266865.1 NP_001253794.1 - 360 162
XM_015114967.2 XP_014970453.1 - 740 162
XM_028843342.1 XP_028699175.1 - 603 162
GenBank mRNA JU332907.1 - - 360 162

View Full Product Details