Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
M1AP
Entrez Gene ID
710210
Gene Name
meiosis 1 associated protein
Gene Aliases
-
Chromosome Location
Chr.13: 34159963 - 34257552 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112712.2 XP_014968198.1 - 622 138
XM_028832358.1 XP_028688191.1 - 622 138

View Full Product Details