Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A13
Entrez Gene ID
693964
Gene Name
solute carrier family 7 member 13
Gene Aliases
-
Chromosome Location
Chr.8: 86626293 - 86643124 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082410.4 XP_001082410.2 - 921 130

View Full Product Details