Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHOXF1
Entrez Gene ID
106995309
Gene Name
Rhox homeobox family member 1
Gene Aliases
EGK_20880
Chromosome Location
Chr.X: 116338458 - 116458150 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028842447.1 XP_028698280.1 - 2260 150
XM_028842448.1 XP_028698281.1 - 3455 150
XM_028842449.1 XP_028698282.1 - 1416 150
XM_028842450.1 XP_028698283.1 - 2355 150
GenBank mRNA JV053164.1 - - 446 150
JV643661.1 - - 484 150

View Full Product Details