Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMTD1
Entrez Gene ID
704152
Gene Name
catechol-O-methyltransferase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 62548881 - 62550933 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261688.1 NP_001248617.1 - 294 132
XM_028853377.1 XP_028709210.1 - 371 132
GenBank mRNA JU332859.1 - - 270 132
JU474259.1 - - 294 132
JV605436.1 - - 296 132

View Full Product Details