Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
717224
Gene Name
DC-STAMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 95999696 - 96015359 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001114929.4 XP_001114929.2 - 1088 103

View Full Product Details