Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT7A
Entrez Gene ID
693419
Gene Name
Wnt family member 7A
Gene Aliases
-
Chromosome Location
Chr.2: 145213924 - 145270951 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265938.2 NP_001252867.1 - 759 80
GenBank mRNA JU320644.1 - - 759 80
JV529645.1 - - 622 80
JV669313.1 - - 789 80

View Full Product Details