Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS3
Entrez Gene ID
715548
Gene Name
dehydrogenase/reductase 3
Gene Aliases
EGK_00247
Chromosome Location
Chr.1: 211831279 - 211883944 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260537.2 NP_001247466.1 - 889 119
GenBank mRNA JU328603.1 - - 665 119
JV044007.1 - - 890 119
JV635508.1 - - 885 119
JV669479.1 - - 843 119

View Full Product Details