Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALYREF
Entrez Gene ID
714852
Gene Name
Aly/REF export factor
Gene Aliases
THOC4
Chromosome Location
Chr.16: 78187103 - 78190724 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261013.1 NP_001247942.1 - 390 86
GenBank mRNA JU473906.1 - - 390 86
JU473907.1 - - 390 86
JV049353.1 - - 266 86
JV581255.1 - - 248 86

View Full Product Details