Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KMT2A
Entrez Gene ID
705511
Gene Name
lysine methyltransferase 2A
Gene Aliases
MLL, all-1
Chromosome Location
Chr.14: 111496876 - 111593102 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115825.2 XP_014971311.2 - 4025 150
XM_015115826.2 XP_014971312.2 - 4025 150
XM_028834035.1 XP_028689868.1 - 4122 150
XM_028834036.1 XP_028689869.1 - 1509 150
GenBank mRNA JU343756.1 - - 3589 150
JU486874.1 - - 3601 150

View Full Product Details