Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC8
Entrez Gene ID
693674
Gene Name
ER membrane protein complex subunit 8
Gene Aliases
COX4NB, EGK_13077
Chromosome Location
Chr.20: 72805778 - 72827904 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265927.1 NP_001252856.1 - 448 121
XM_015126712.2 XP_014982198.1 - 539 121
GenBank mRNA JU320694.1 - - 461 121
JU472808.1 - - 437 121
JU472809.1 - - 437 121
JV045890.1 - - 448 121

View Full Product Details