Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC2
Entrez Gene ID
710469
Gene Name
WAP four-disulfide core domain 2
Gene Aliases
EGK_02361
Chromosome Location
Chr.10: 80700954 - 80712633 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU325708.1 - - 121 144
JV045901.1 - - 251 144

View Full Product Details