Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN5
Entrez Gene ID
697372
Gene Name
fibulin 5
Gene Aliases
-
Chromosome Location
Chr.7: 154013267 - 154094084 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260534.1 NP_001247463.1 - 1452 82
XM_015144314.2 XP_014999800.1 - 1338 82
GenBank mRNA JV045929.1 - - 1452 82

View Full Product Details